Novus Biologicals
Manufacturer Code:NBP297837AF700
Catalog # NB103659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RNGTT fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP1AmRNA-capping enzyme hCAP HCE1DKFZp686J2031 HCEHCAP1 RNA guanylyltransferase and 5'-phosphatase; GTase; GTP--RNA guanylyltransferase; HCAP1; HCE; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; TPase
Gene Aliases: CAP1A; hCAP; HCE; HCE1; RNGTT
UniProt ID: (Human) O60942
Entrez Gene ID: (Human) 8732
Molecular Function:
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.