Invitrogen
Manufacturer Code:PA520961
Catalog # PIPA520961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the carboxy terminus of human RNF8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ActA binding protein 2; ActA-interacting protein 37; AIP37; C3HC4-type zinc finger protein; C3HC4-type zinc finger protein E3 ubiquitin-protein ligase RNF8 UBC13/UEV-interacting ring finger protein ring finger protein (C3HC4 type) 8; E3 ubiquitin-protein ligase RNF8; hRNF8; LaXp180; ring finger protein (C3HC4 type) 8; RING finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; UBC13/UEV-interacting ring finger protein
Gene Aliases: 3830404E21Rik; AIP37; hRNF8; KIAA0646; RNF8
UniProt ID: (Human) O76064, (Rat) Q4KLN8, (Mouse) Q8VC56
Entrez Gene ID: (Human) 9025, (Rat) 361815, (Mouse) 58230
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.