Invitrogen
Manufacturer Code:PA518132
Catalog # PIPA518132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GEPGFFVTGDRAG) corresponding to the N-terminus amino acids of RNF8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C3HC4-type zinc finger protein; E3 ubiquitin-protein ligase RNF8; hRNF8; ring finger protein (C3HC4 type) 8; RING finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; RNF8 ring finger protein 8 ring finger protein (C3HC4 type) 8 KIAA0646 FLJ12013 C3HC4-type zinc finger protein OTTHUMP00000039684 UBC13/UEV-interacting ring finger protein; UBC13/UEV-interacting ring finger protein
Gene Aliases: hRNF8; KIAA0646; RNF8
UniProt ID: (Human) O76064
Entrez Gene ID: (Human) 9025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.