Novus Biologicals
Manufacturer Code:NBP257378
Catalog # NBP257378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CEVTVGRGFGVTYQLVSKICPLMISRNHCVLKQNPEGQWTIMDNKSLNGVWLNRARLEPLRVYSIHQGDYIQLGVPLENKENAEYEYEVTEEDWETIYPCLSPKNDQMI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C3HC4-type zinc finger protein; C3HC4-type zinc finger protein EC 6.3.2 EC 6.3.2.- FLJ12013 KIAA0646UBC13/UEV-interacting ring finger protein ring finger protein (C3HC4 type) 8 ring finger protein 8E3 ubiquitin-protein ligase RNF8; E3 ubiquitin-protein ligase RNF8; hRNF8; ring finger protein (C3HC4 type) 8; RING finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; UBC13/UEV-interacting ring finger protein
Gene Aliases: hRNF8; KIAA0646; RNF8
UniProt ID: (Human) O76064
Entrez Gene ID: (Human) 9025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.