R&D Systems
Manufacturer Code:AF7114SP
Catalog # AF7114SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human RNF8 Met1-Val124 Accession # O76064 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ActA Binding Protein 2 C3HC4-type zinc finger protein EC 6.3.2 EC 6.3.2.- FLJ12013 KIAA0646UBC13/UEV-interacting ring finger protein ring finger protein (C3HC4 type) 8 ring finger protein 8E3 ubiquitin-protein ligase RNF8; C3HC4-type zinc finger protein; E3 ubiquitin-protein ligase RNF8; hRNF8; ring finger protein (C3HC4 type) 8; RING finger protein 8; ring finger protein 8, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase RNF8; UBC13/UEV-interacting ring finger protein
Gene Aliases: hRNF8; KIAA0646; RNF8
UniProt ID: (Human) O76064
Entrez Gene ID: (Human) 9025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.