Proteintech
Manufacturer Code:119051AP150UL
Catalog # 50-172-7549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF7 Fusion Protein Ag2553 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CKBBP1; CKBBP1 CKII beta binding protein 1 Rbx2 Regulator of cullins 2 RING box protein 2 ring finger protein 7 RNF7 ROC2 SAG; CKII beta-binding protein 1; Rbx2; Regulator of cullins 2; RING finger protein 7; RING-box protein 2; Sensitive to apoptosis gene protein; sensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2; zinc RING finger protein SAG
Gene Aliases: CKBBP1; RBX2; RNF7; ROC2; SAG
UniProt ID: (Human) Q9UBF6, (Mouse) Q9WTZ1
Entrez Gene ID: (Human) 9616, (Mouse) 19823, (Rat) 300948
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.