Novus Biologicals
Manufacturer Code:NB100583
Catalog # NB100583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 275 and the C-terminus (residue 317) of human Ring Finger Protein 41 using the number given in entry NP_005776.1 (GeneID 10193). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase NRDP1; E3 ubiquitin-protein ligase NRDP1 EC 6.3.2 EC 6.3.2.- FLRF MGC45228 NRDP1 ring finger protein 41neuregulin receptor degradation protein-1 SBBI03; fetal liver ring finger; neuregulin receptor degradation protein-1; RING finger protein 41; ring finger protein 41, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase NRDP1
Gene Aliases: FLRF; NRDP1; RNF41; SBBI03
UniProt ID: (Human) Q9H4P4
Entrez Gene ID: (Human) 10193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.