R&D Systems
Manufacturer Code:AF7964SP
Catalog # AF7964SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E.coli</em>-derived recombinant human RNF4 Met1-Ile190 Accession # P78317 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin ligase RNF4; E3 ubiquitin ligase RNF4 EC 6.3.2.- Protein SNURF RES4-26 ring finger protein 4SNURFSLX5 SLX5 Small nuclear ring finger protein SNURF; E3 ubiquitin-protein ligase RNF4; Protein SNURF; RING finger protein 4; RING-type E3 ubiquitin transferase RNF4; Small nuclear ring finger protein
Gene Aliases: RES4-26; RNF4; SLX5; SNURF
UniProt ID: (Human) P78317
Entrez Gene ID: (Human) 6047
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.