Abnova
Manufacturer Code:H00006047A01
Catalog # 89-105-339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 107 a.a. &bsim 190 a.a) partial recombinant protein with GST tag.,RNF4 (NP_002929 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant RNF4.
For Research Use Only
Protein Aliases: E3 ubiquitin ligase RNF4; E3 ubiquitin-protein ligase RNF4; Protein SNURF; RES4-26; RING finger protein 4; RING-type E3 ubiquitin transferase RNF4; Small nuclear ring finger protein; SNURF
Gene Aliases: RES4-26; RNF4; SLX5; SNURF
UniProt ID: (Human) P78317
Entrez Gene ID: (Human) 6047
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.