Proteintech
Manufacturer Code:178101AP150UL
Catalog # 50-100-2685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF4 Fusion Protein Ag12115 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin ligase RNF4; E3 ubiquitin ligase RNF4 Protein SNURF RES4 26 ring finger protein 4 RNF4 SNURF; E3 ubiquitin-protein ligase RNF4; Protein SNURF; RING finger protein 4; RING-type E3 ubiquitin transferase RNF4; Small nuclear ring finger protein; SNURF
Gene Aliases: AU018689; Gtrgeo8; RES4-26; RNF4; SLX5; SNURF
UniProt ID: (Human) P78317, (Rat) O88846, (Mouse) Q9QZS2
Entrez Gene ID: (Human) 6047, (Rat) 29274, (Mouse) 19822
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.