Invitrogen
Manufacturer Code:PA5113296
Catalog # PIPA5113296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human E3 ubiquitin-protein ligase RNF34 protein (1-220AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW061037 AW536122 BC004042 C88279 CARP1 CARP-1 Caspase regulator CARP1 Caspases-8 and -10-associated RING finger protein 1 E3 ubiquitin-protein ligase RNF34 FLJ21786 FYVE-RING finger protein Momo hRFI Human RING finger homologous to inhibitor of apoptosis protein Momo phafin 1 Phafin-1 RFI RIF RIFF RING finger protein 34 ring finger protein 34 E3 ubiquitin protein ligase RING finger protein MOMO RING finger protein RIFF RING-type E3 ubiquitin transferase RNF34 RNF34; CARP-1; Caspase regulator CARP1; Caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; RING finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34
Gene Aliases: CARP-1; CARP1; hRFI; RFI; RIF; RIFF; RNF34
UniProt ID: (Human) Q969K3
Entrez Gene ID: (Human) 80196
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.