Proteintech
Manufacturer Code:106291AP150UL
Catalog # 50-172-6460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF34 Fusion Protein Ag0982 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARP 1 Caspase regulator CARP1 FYVE RING finger protein Momo hRFI RFI RIF RIFF ring finger protein 34 RING finger protein RIFF RNF34; CARP-1; Caspase regulator CARP1; Caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; phafin 1; Phafin-1; RING finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34
Gene Aliases: AW061037; AW536122; BC004042; C88279; CARP-1; CARP1; hRFI; Momo; RFI; RIF; RIFF; RNF34
UniProt ID: (Human) Q969K3, (Mouse) Q99KR6, (Rat) Q6AYH3
Entrez Gene ID: (Human) 80196, (Mouse) 80751, (Rat) 282845
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.