Abnova
Manufacturer Code:H00080196K
Catalog # 89-208-704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RNF34 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RNF34 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CARP-1; Caspase regulator CARP1; Caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786 RFI RIF RIFF; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; RING finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34
Gene Aliases: CARP-1; CARP1; hRFI; RFI; RIF; RIFF; RNF34
UniProt ID: (Human) Q969K3
Entrez Gene ID: (Human) 80196
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.