Novus Biologicals
Manufacturer Code:NB10055427
Catalog # NB10055427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KTAAVLKWNRE corresponding to C-Terminus according to NP_065965.4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ALK lymphoma oligomerization partner on chromosome 17; ALK lymphoma oligomerization partner on chromosome 17 C17orf27 chromosome 17 open reading frame 27 DKFZp762N1115 FLJ13051 KIAA1554MGC46622 KIAA1618 MGC9929 NET57 protein ALO17 ring finger protein 213; E3 ubiquitin-protein ligase RNF213; Mysterin; RING finger protein 213; RING-type E3 ubiquitin transferase RNF213
Gene Aliases: ALO17; C17orf27; KIAA1554; KIAA1618; MYMY2; MYSTR; NET57; RNF213
UniProt ID: (Human) Q63HN8
Entrez Gene ID: (Human) 57674
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.