Novus Biologicals
Manufacturer Code:NBP155083
Catalog # NBP155083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RNF212(ring finger protein 212) The peptide sequence was selected from the middle region of RNF212. Peptide sequence LCKKYSRETSQILEFQEKHRKRLLAFYREKISRLEESLRKSVLQIEQLQS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ38841 FLJ42587 hypothetical protein LOC285498 MGC120227 MGC120228 ring finger protein 212 ZHP3; Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212
Gene Aliases: RNF212; ZHP3
UniProt ID: (Human) Q495C1
Entrez Gene ID: (Human) 285498
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.