Proteintech
Manufacturer Code:211781AP150UL
Catalog # 50-173-3301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF212 Fusion Protein Ag15457 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Probable E3 SUMO-protein ligase RNF212; Probable E3 SUMO-protein transferase RNF212; RING finger protein 212; ring finger protein 212 RNF212 ZHP3
Gene Aliases: EG330157; F830003B07; RNF212; ZHP3
UniProt ID: (Human) Q495C1
Entrez Gene ID: (Human) 285498, (Mouse) 671564, (Rat) 689721
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.