Invitrogen
Manufacturer Code:PA572027
Catalog # PIPA572027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 528-555 amino acids from the C-terminal region of human RNF207 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RING finger protein 207; RING finger protein 207 RNF207 C1orf188
Gene Aliases: C1orf188; D330010C22Rik; Gm143; RNF207
UniProt ID: (Human) Q6ZRF8, (Mouse) Q3V3A7
Entrez Gene ID: (Human) 388591, (Mouse) 433809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.