Novus Biologicals
Manufacturer Code:NB1002243
Catalog # NB1002243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human Ring Finger 20 using the numbering given in entry NP_062538.5 (GeneID 56254). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRE1 BRE1 E3 ubiquitin ligase homolog BRE1-A BRE1AMGC129667 E3 ubiquitin-protein ligase BRE1A EC 6.3.2 EC 6.3.2.- FLJ11189 FLJ20382 hBRE1MGC129668 homolog of S. cerevisiae BRE1 KAIA2779 ring finger protein 20KIAA2779; BRE1 E3 ubiquitin ligase homolog; BRE1-A; E3 ubiquitin-protein ligase BRE1A; homolog of S. cerevisiae BRE1; RING finger protein 20; RING-type E3 ubiquitin transferase BRE1A
Gene Aliases: BRE1; BRE1A; hBRE1; RNF20
UniProt ID: (Human) Q5VTR2
Entrez Gene ID: (Human) 56254
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.