Abnova
Manufacturer Code:H00056254B01
Catalog # 89-007-928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF20 (NP_062538.5 1 a.a. ~ 975 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human RNF20 protein.
For Research Use Only
Protein Aliases: BRE1; BRE1 E3 ubiquitin ligase homolog; BRE1-A; BRE1A; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; hBRE1; homolog of S. cerevisiae BRE1; KIAA2779; MGC129667; MGC129668; RING finger protein 20; RING-type E3 ubiquitin transferase BRE1A
Gene Aliases: BRE1; BRE1A; hBRE1; RNF20
UniProt ID: (Human) Q5VTR2
Entrez Gene ID: (Human) 56254
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.