Proteintech
Manufacturer Code:216251AP150UL
Catalog # 50-173-3519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF20 Fusion Protein Ag16066 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRE1 BRE1 A BRE1A hBRE1 KIAA2779 ring finger protein 20 RNF20; BRE1 E3 ubiquitin ligase homolog; BRE1-A; E3 ubiquitin-protein ligase BRE1A; homolog of S. cerevisiae BRE1; RING finger protein 20; RING-type E3 ubiquitin transferase BRE1A
Gene Aliases: 4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; Kiaa4116; mKIAA4116; RNF20
UniProt ID: (Human) Q5VTR2, (Mouse) Q5DTM8
Entrez Gene ID: (Human) 56254, (Mouse) 109331, (Rat) 313216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.