Invitrogen
Manufacturer Code:PA540858
Catalog # PIPA540858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RNF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RING2; HIP2-interacting protein 3; Huntingtin-interacting protein 2-interacting protein 3; Protein DinG; RING finger protein 1B; RING finger protein 2; Ring finger protein 2 BAP1 DING BAP-1 HIPI3 RING2 RING1B; RING finger protein BAP-1; RING-type E3 ubiquitin transferase RING2; RING1b; rnf2
Gene Aliases: AI326319; AI450156; AU019207; BAP-1; BAP1; DING; fa19h05; HIPI3; RING1B; RING2; RNF2; wu:fa19h05; wz4556; wz4566
UniProt ID: (Human) Q99496, (Rat) Q4KLY4, (Mouse) Q9CQJ4
Entrez Gene ID: (Human) 6045, (Rabbit) 100344273, (Bovine) 540090, (Dog) 480042, (Horse) 100056141, (Guinea pig) 100717673, (Rat) 304850, (Mouse) 19821, (Zebrafish) 327035
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.