Novus Biologicals
Manufacturer Code:NBP170695
Catalog # NBP170695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RNF165(ring finger protein 165) The peptide sequence was selected from the N terminal of RNF165. Peptide sequence MVLVHVGYLVLPVFGSVRNRGAPFQRSQHPHATSCRHFHLGPPQPQQLAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARKL2 ring finger protein 165; E3 ubiquitin-protein ligase RNF165; RING finger protein 165
Gene Aliases: ARKL2; RNF111L2; RNF165
UniProt ID: (Human) Q6ZSG1
Entrez Gene ID: (Human) 494470
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.