Proteintech
Manufacturer Code:112851AP150UL
Catalog # 50-172-7031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNF156 Fusion Protein Ag1808 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase MGRN1; KIAA0544 mahogunin ring finger 1 MGRN1 RING finger protein 156 RNF156; mahoganoid; mahogunin ring finger 1, E3 ubiquitin protein ligase; Mahogunin RING finger protein 1; mahogunin, ring finger 1; probable E3 ubiquitin-protein ligase MGRN1; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1
Gene Aliases: 2610042J20Rik; KIAA0544; md; MGRN1; mKIAA0544; nc; RGD1311862; RNF156
UniProt ID: (Human) O60291, (Rat) Q5XIQ4, (Mouse) Q9D074
Entrez Gene ID: (Human) 23295, (Rat) 302938, (Mouse) 17237
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.