Invitrogen
Manufacturer Code:PA540866
Catalog # PIPA540866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RNF10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RING finger protein 10; Ring finger protein 10 KIAA0262 MGC126758 MGC126764 RIE2; Sid 2705
Gene Aliases: AA675014; KIAA0262; mKIAA0262; RIE2; RNF10; Sid2705
UniProt ID: (Human) Q8N5U6, (Bovine) Q08E13, (Mouse) Q3UIW5, (Rat) Q5XI59
Entrez Gene ID: (Human) 9921, (Dog) 477514, (Bovine) 520757, (Horse) 100050260, (Mouse) 50849, (Rat) 288710, (Guinea pig) 100727163
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.