Novus Biologicals
Manufacturer Code:NBP238672
Catalog # NBP238672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SPCYYFYQAEDGQHMFLHPVNVRCLVREYGSLERSPEKISATVVEIAGYSMSEDVRQRHRYLSHLPLTCEFSICELALQPPVV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0262RIE2MGC126758 MGC126764 ring finger protein 10; RING finger protein 10
Gene Aliases: KIAA0262; RIE2; RNF10
UniProt ID: (Human) Q8N5U6
Entrez Gene ID: (Human) 9921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.