Invitrogen
Manufacturer Code:PA520667
Catalog # PIPA520667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the center of human RNAse H2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGS4; AGS4 JUNB RNASEHI RNHIA RNHL RNase H(35) RNase H2 subunit A RNase HI large subunit aicardi-Goutieres syndrome 4 protein ribonuclease H2 subunit A ribonuclease H2 large subunit ribonuclease HI large subunit ribonuclease HI subunit A ribonuclease HI large subunit; Aicardi-Goutieres syndrome 4 protein; Ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease H2, subunit A; Ribonuclease HI large subunit; Ribonuclease HI subunit A; ribonuclease HI, large subunit; RNase H(35); RNase H2 subunit A; RNase HI large subunit
Gene Aliases: 2400006P09Rik; AGS4; JUNB; RNASEH2A; RNASEHI; RNHIA; RNHL
UniProt ID: (Human) O75792, (Mouse) Q9CWY8, (Rat) Q5U209
Entrez Gene ID: (Human) 10535, (Mouse) 69724, (Rat) 364974
Molecular Function:
RNA binding protein
endoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.