Proteintech
Manufacturer Code:165181AP150UL
Catalog # 50-173-1002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNASEH2C Fusion Protein Ag9739 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGS3; AGS3 AYP1 Ribonuclease H2 subunit C ribonuclease H2 subunit C Ribonuclease HI subunit C RNase H1 small subunit RNase H2 subunit C RNASEH2C; Aicardi-Goutieres syndrome 3 protein; Ribonuclease H2 subunit C; ribonuclease H2, subunit C; Ribonuclease HI subunit C; RNase H1 small subunit; RNase H2 subunit C
Gene Aliases: AGS3; AYP1; RNASEH2C
UniProt ID: (Human) Q8TDP1
Entrez Gene ID: (Human) 84153
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.