Invitrogen
Manufacturer Code:PA524708
Catalog # PIPA524708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 130-157 amino acids from the central region of human RNASEH2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGS2; AGS2 DLEU8 Aicardi-Goutieres syndrome 2 protein RNase H2 subunit B deleted in lymphocytic leukemia 8 ribonuclease H2 subunit B ribonuclease HI subunit B; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; Ribonuclease H2 subunit B; ribonuclease H2, subunit B; Ribonuclease HI subunit B; RNase H2 subunit B
Gene Aliases: AGS2; DLEU8; RNASEH2B
UniProt ID: (Human) Q5TBB1
Entrez Gene ID: (Human) 79621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.