Proteintech
Manufacturer Code:175271AP150UL
Catalog # 50-173-1626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNASEH2B Fusion Protein Ag11189 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGS2; AGS2 DLEU8 Ribonuclease H2 subunit B ribonuclease H2 subunit B Ribonuclease HI subunit B RNase H2 subunit B RNASEH2B; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; Deleted in lymphocytic leukemia 8 homolog; Ribonuclease H2 subunit B; ribonuclease H2, subunit B; Ribonuclease HI subunit B; RNase H2 subunit B
Gene Aliases: 1110019N06Rik; 2610207P08Rik; AGS2; DLEU8; RNASEH2B
UniProt ID: (Human) Q5TBB1, (Mouse) Q80ZV0, (Rat) Q5XI96
Entrez Gene ID: (Human) 79621, (Mouse) 67153, (Rat) 361056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.