Novus Biologicals
Manufacturer Code:NBP158234
Catalog # NBP158234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RNASEH2A(ribonuclease H2 subunit A) The peptide sequence was selected from the C terminal of RNASEH2A. Peptide sequence EKEAEDVIWEDSASENQEGLRKITSYFLNEGSQARPRSSHRYFLERGLES. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGS4; AGS4RNase H2 subunit A Aicardi-Goutieres syndrome 4 Aicardi-Goutieres syndrome 4 protein JUNB ribonuclease H2 subunit A ribonuclease H2 large subunit ribonuclease H2 subunit A Ribonuclease HI large subunit Ribonuclease HI subunit A ribonuclease HI large subunit RNase H(35) RNASEHIEC 3.1.26.4 RNHIARNase HI large subunit RNHL; Aicardi-Goutieres syndrome 4 protein; Ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease H2, subunit A; Ribonuclease HI large subunit; Ribonuclease HI subunit A; ribonuclease HI, large subunit; RNase H(35); RNase H2 subunit A; RNase HI large subunit
Gene Aliases: AGS4; JUNB; RNASEH2A; RNASEHI; RNHIA; RNHL
UniProt ID: (Human) O75792
Entrez Gene ID: (Human) 10535
Molecular Function:
RNA binding protein
endoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.