Proteintech
Manufacturer Code:156061AP150UL
Catalog # 50-554-698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNASEH1 Fusion Protein Ag7990 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H1RNA Ribonuclease H type II ribonuclease H1 RNase H1 RNASEH1 RNH1; Ribonuclease H type II; Ribonuclease H1; RNase H1
Gene Aliases: H1RNA; PEOB2; RNASEH1; RNH1
UniProt ID: (Human) O60930, (Rat) Q5BK46
Entrez Gene ID: (Human) 246243, (Mouse) 19819, (Rat) 298933
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.