OriGene
Manufacturer Code:TA811245
Catalog # 50-168-3932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F12 |
Immunogen | Full length human recombit protein of human RNASE9 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HEL128 RAK1 h461; Inactive ribonuclease-like protein 9; ribonuclear enzyme; ribonuclease A K1; ribonuclease, RNase A family, 9 (non-active)
Gene Aliases: h461; HEL128; RAK1; RNASE9
UniProt ID: (Human) P60153
Entrez Gene ID: (Human) 390443
Molecular Function: RNA binding protein endoribonuclease enzyme modulator hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.