Invitrogen
Manufacturer Code:MA524591
Catalog # PIMA524591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0224 |
Immunogen | Recombit protein corresponding to Human RNASE7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ribonuclease 7; ribonuclease, RNase A family, 7; RNase 7; SAP-2; Skin-derived antimicrobial protein 2
Gene Aliases: RAE1; RNASE7; UNQ2516/PRO6006
UniProt ID: (Human) Q9H1E1
Entrez Gene ID: (Human) 84659
Molecular Function:
RNA binding protein
endoribonuclease
enzyme modulator
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.