Abnova
Manufacturer Code:H00023543M01
Catalog # 89-014-289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G3 |
Immunogen | RBM9 (AAH13115 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant RBM9.
For Research Use Only
Protein Aliases: dJ106I20.3; Fox-1 homolog B; fox-1 homologue; Fox-2; FOX2; fxh; Hexaribonucleotide-binding protein 2; HNRBP2; HRNBP2; Repressor of tamoxifen transcriptional activity; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA-binding motif protein 9; RNA-binding protein 9; RTA
Gene Aliases: dJ106I20.3; Fox-2; FOX2; fxh; HNRBP2; HRNBP2; RBFOX2; RBM9; RTA
UniProt ID: (Human) O43251
Entrez Gene ID: (Human) 23543
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.