Novus Biologicals
Manufacturer Code:NB200598R
Catalog # NB200598R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H8 |
Immunogen | 10 repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser 5 YSPTSpPS (Human). [UniProt# P24928] |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II largest subunit RNA polymerase II 220 kdsubunit DNA-directed RNA polymerase II subunit A DNA-directed RNA polymerase II subunit RPB1 DNA-directed RNA polymerase III largest subunit EC 2.7.7.48 EC 2.7.7.6 hRPB220 hsRPB1 MGC75453 POLR2 POLRA polymerase (RNA) II (DNA directed) polypeptide A (220kD) polymerase (RNA) II (DNA directed) polypeptide A 220kDa RNA Pol 2 RNA Polymerase 2 RNA polymerase II subunit B1 RNA-directed RNA polymerase II subunit RPB1 RNAPII RPB1 RPBh1 RpIILS RPO2 RPOL2; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1
Gene Aliases: hRPB220; hsRPB1; POLR2; POLR2A; POLRA; RPB1; RPBh1; RpIILS; RPO2; RPOL2
UniProt ID: (Human) P24928
Entrez Gene ID: (Human) 5430
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.