Invitrogen
Manufacturer Code:PA542857
Catalog # PIPA542857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RMI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BLAP75; BLM-associated polypeptide, 75 kDa; BLM-associated protein 75 kDa; BLM-associated protein of 75 kDa; FAAP75; homolog of yeast RecQ-mediated genome instability 1 (RMI1); RecQ mediated genome instability 1; RecQ-mediated genome instability protein 1; recQ-mediated genome instability protein-like protein; RMI1 RecQ mediated genome instability 1 homolog (S. cerevisiae) BLAP75 C9orf76 FLJ12888 RP11-346I8.1; RMI1, RecQ mediated genome instability 1, homolog
Gene Aliases: 4932432N11Rik; BLAP75; C79893; C9orf76; FAAP75; RGD1310671; RMI1
UniProt ID: (Human) Q6P1Q4, (Bovine) A4IF98, (Mouse) Q9D4G9
Entrez Gene ID: (Rabbit) 100352297, (Human) 80010, (Bovine) 614063, (Dog) 100684392, (Horse) 100052791, (Guinea pig) 100724405, (Rat) 306734, (Mouse) 74386
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.