Invitrogen
Manufacturer Code:PA5110300
Catalog # PIPA5110300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human RMI1 (NP_079221.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4932432N11Rik BLAP75 BLM-associated protein of 75 kDa BOS_9139 C79893 C9orf76 FAAP75 FLJ12888 RecQ-mediated genome instability protein 1 RGD1310671 RMI1 RP11-346I8.1; BLAP75; BLM-associated polypeptide, 75 kDa; BLM-associated protein 75 kDa; BLM-associated protein of 75 kDa; FAAP75; homolog of yeast RecQ-mediated genome instability 1 (RMI1); RecQ-mediated genome instability protein 1; RMI1, RecQ mediated genome instability 1, homolog
Gene Aliases: 4932432N11Rik; BLAP75; C79893; C9orf76; FAAP75; RMI1
UniProt ID: (Human) Q6P1Q4, (Mouse) Q9D4G9
Entrez Gene ID: (Human) 80010, (Mouse) 74386
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.