Proteintech
Manufacturer Code:146301AP150UL
Catalog # 50-172-9599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RMI1 Fusion Protein Ag6208 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BLAP75; BLAP75 C9orf76 FAAP75 RMI1 RP11 346I8.1; BLM-associated polypeptide, 75 kDa; BLM-associated protein 75 kDa; BLM-associated protein of 75 kDa; FAAP75; homolog of yeast RecQ-mediated genome instability 1 (RMI1); RecQ-mediated genome instability protein 1; RMI1, RecQ mediated genome instability 1, homolog
Gene Aliases: BLAP75; C9orf76; FAAP75; RMI1
UniProt ID: (Human) Q6P1Q4
Entrez Gene ID: (Human) 80010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.