Invitrogen
Manufacturer Code:MA528632
Catalog # PIMA528632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EM-53 |
Immunogen | Murine Rltpr (amino acids 1147-1397) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Capping protein regulator and myosin 1 linker 2; Capping protein, Arp2/3 and myosin-I linker protein 2; F-actin-uncapping protein RLTPR; leucine rich repeat containing 16C; Leucine-rich repeat-containing protein 16C; RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein; RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein
Gene Aliases: CARMIL2; CARMIL2b; CG1399-PB; D130029J02Rik; Gm585; LRRC16C; RLTPR
UniProt ID: (Human) Q6F5E8
Entrez Gene ID: (Human) 146206, (Mouse) 234695
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.