Proteintech
Manufacturer Code:213941AP150UL
Catalog # 50-173-3403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RLBP1L2 Fusion Protein Ag16061 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf212 C6orf213 Clavesin 2 CLVS2 RLBP1L2; clathrin vesicle-associated Sec14 protein 2; Clavesin-2; clavesin-2 {ECO:0000303|PubMed:19651769}; clvs2 {ECO:0000303|PubMed:19651769}; retinaldehyde binding protein 1-like 2; Retinaldehyde-binding protein 1-like 2; Retinaldehyde-binding protein 1-like protein 2
Gene Aliases: A330019N05Rik; bA160A10.4; C6orf212; C6orf213; Clavesin-2; CLVS2; RGD1306801; RLBP1L2
UniProt ID: (Human) Q5SYC1, (Mouse) Q8BG92
Entrez Gene ID: (Human) 134829, (Mouse) 215890, (Rat) 361459
Molecular Function:
dehydrogenase
oxidoreductase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.