OriGene
Manufacturer Code:TA503151
Catalog # 50-167-4118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G2 |
Immunogen | Full length human recombit protein of human RLBP1 produced in HEK293 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; Retinaldehyde-binding protein 1
Gene Aliases: CRALBP; RLBP1
UniProt ID: (Human) P12271
Entrez Gene ID: (Human) 6017
Molecular Function: dehydrogenase oxidoreductase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.