Proteintech
Manufacturer Code:153561AP150UL
Catalog # 50-173-0173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RLBP1 Fusion Protein Ag7602 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP RLBP1; Retinaldehyde-binding protein 1
Gene Aliases: 3110056M11Rik; CRALBP; RLBP1
UniProt ID: (Human) P12271, (Mouse) Q9Z275
Entrez Gene ID: (Human) 6017, (Mouse) 19771, (Rat) 293049
Molecular Function: dehydrogenase oxidoreductase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.