Novus Biologicals
Manufacturer Code:NBP29476920UL
Catalog # NB024055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PRDM2 (NP_036363.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.43 GATA-3 binding protein G3B GATA-3-binding protein G3B KMT8PR domain-containing protein 2 Lysine N-methyltransferase 8 MTB-ZFHUMHOXY1 PR domain containing 2 with ZNF domain PR domain zinc finger protein 2 retinoblastoma protein-binding zinc finger protein Retinoblastoma protein-interacting zinc finger protein RIZ1 RIZ2 RIZMTE-binding protein Zinc finger protein RIZ zinc-finger DNA-binding protein; GATA-3 binding protein G3B; GATA-3-binding protein G3B; Lysine N-methyltransferase 8; MTB-ZF; MTE-binding protein; PR domain containing 2, with ZNF domain; PR domain zinc finger protein 2; PR domain-containing protein 2; retinoblastoma protein-binding zinc finger protein; Retinoblastoma protein-interacting zinc finger protein; Zinc finger protein RIZ; zinc-finger DNA-binding protein
Gene Aliases: HUMHOXY1; KMT8; MTB-ZF; PRDM2; RIZ; RIZ1; RIZ2
UniProt ID: (Human) Q13029
Entrez Gene ID: (Human) 7799
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.