Novus Biologicals
Manufacturer Code:NBP273912AF700
Catalog # N273912A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B3 |
Immunogen | RIPK3/RIP3 Antibody (OTI1B3) [Alexa Fluor 700] was made to a recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3/RIP3 (NP_006862) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.11.1 Receptor Interacting Serine/Threonine Kinase 3 Receptor-interacting protein 3 receptor-interacting serine/threonine-protein kinase 3 receptor-interacting serine-threonine kinase 3 RIP3 RIP-3 RIP-like protein kinase 3; receptor interacting protein 3; Receptor-interacting protein 3; receptor-interacting serine-threonine kinase 3; Receptor-interacting serine/threonine-protein kinase 3; RIP-3; RIP-like protein kinase 3
Gene Aliases: RIP3; RIPK3
UniProt ID: (Human) Q9Y572
Entrez Gene ID: (Human) 11035
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.