OriGene
Manufacturer Code:CF803093
Catalog # 50-166-6324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 233-518 of human RIPK3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: receptor interacting protein 3; Receptor-interacting protein 3; receptor-interacting serine-threonine kinase 3; Receptor-interacting serine/threonine-protein kinase 3; RIP-3; RIP-like protein kinase 3
Gene Aliases: RIP3; RIPK3
UniProt ID: (Human) Q9Y572
Entrez Gene ID: (Human) 11035
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.