R&D Systems
Manufacturer Code:MAB103871100
Catalog # MAB103871100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1018218 |
Immunogen | <i>E. coli</i>-derived human RIPK2/RIP2 Met1-Met540 Accession &num O43353 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARD3 CARD-containing IL-1 beta ICE-kinase CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase CARD-containing interleukin-1 beta-converting enzyme-associated kinase CARDIAKCCK EC 2.7.11 EC 2.7.11.1 GIG30 growth-inhibiting gene 30 receptor interacting protein 2 receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase Receptor-interacting protein 2 receptor-interacting serine/threonine-protein kinase 2 receptor-interacting serine-threonine kinase 2 RICK RICKGIG30 RIP2 RIP-2 RIP2CARD-carrying kinase RIP-like-interacting CLARP kinase; growth-inhibiting gene 30; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RIP-2; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2
Gene Aliases: CARD3; CARDIAK; CCK; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092
UniProt ID: (Human) O43353
Entrez Gene ID: (Human) 8767
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.