Novus Biologicals
Manufacturer Code:NBP186630
Catalog # NBP186630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LPAPQDNDFLSRKAQDCYFMKLHHCPGNHSWDSTISGSQRAAFCDHKTTPCSSAIINPLSTAGNSERLQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARD3 CARD-containing IL-1 beta ICE-kinase CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase CARD-containing interleukin-1 beta-converting enzyme-associated kinase CARDIAKCCK EC 2.7.11 EC 2.7.11.1 growth-inhibiting gene 30 receptor interacting protein 2 receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase Receptor-interacting protein 2 receptor-interacting serine/threonine-protein kinase 2 receptor-interacting serine-threonine kinase 2 RICKGIG30 RIP-2 RIP2CARD-carrying kinase RIP-like-interacting CLARP kinase; growth-inhibiting gene 30; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RIP-2; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2
Gene Aliases: CARD3; CARDIAK; CCK; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092
UniProt ID: (Human) O43353
Entrez Gene ID: (Human) 8767
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.