Abnova
Manufacturer Code:PAB18782
Catalog # 89-120-339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 15 amino acids at C-terminus of human RIPK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RIPK2.
For Research Use Only
Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARD3; CARDIAK; CCK; GIG30; growth-inhibiting gene 30; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RICK; RIP-2; RIP-like-interacting CLARP kinase; RIP2; Tyrosine-protein kinase RIPK2
Gene Aliases: CARD3; CARDIAK; CCK; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092
UniProt ID: (Human) O43353
Entrez Gene ID: (Human) 8767
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.