Invitrogen
Manufacturer Code:MA517221
Catalog # PIMA517221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AF28D3 |
Immunogen | Recombit human protein purified from E.coli (His-RIP2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; CARD3 CARDIAK CCK GIG30 RICK RIP2 CARD-carrying kinase CARD-containing IL-1 beta ICE-kinase CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase CARD-containing interleukin-1 beta-converting enzyme-associated kinase RIP-2; growth-inhibiting gene 30; receptor (TNFRSF)-interacting serine-threonine kinase 2; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RIP-2; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2
Gene Aliases: 2210420D18Rik; CARD3; CARDIAK; CCK; D4Bwg0615e; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092
UniProt ID: (Human) O43353, (Mouse) P58801
Entrez Gene ID: (Human) 8767, (Rat) 362491, (Mouse) 192656
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.