Invitrogen
Manufacturer Code:PA519933
Catalog # PIPA519933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to amino acids 508 to 522 of human origin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD-carrying kinase; CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase; CARD-containing interleukin-1 beta-converting enzyme-associated kinase; growth-inhibiting gene 30; receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase; Receptor-interacting protein 2; receptor-interacting serine-threonine kinase 2; Receptor-interacting serine/threonine-protein kinase 2; RIP-2; RIP-like-interacting CLARP kinase; Tyrosine-protein kinase RIPK2; WUGSC:H&lowbarRG437L15.1 CARD3 CARDIAK CCK GIG30 RICK RIP2 CARD-carrying kinase CARD-containing IL-1 beta ICE-kinase CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase CARD-containing interleukin-1 beta-converting enzyme-associated kinase RIP-2 RIP-like-interacting CLARP kinase growth-inhibiting gene 30 receptor interacting protein 2 receptor-interacting protein (RIP)-like interacting caspase-like apoptosis regulatory protein (CLARP) kinase receptor-interactin
Gene Aliases: CARD3; CARDIAK; CCK; GIG30; RICK; RIP2; RIPK2; UNQ277/PRO314/PRO34092
UniProt ID: (Human) O43353
Entrez Gene ID: (Human) 8767
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.